Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
Potential calcium-dependent cell-adhesion protein. May be involved in the establishment and maintenance of specific neuronal connections in the brain.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: KIAA0345-like 1; ortholog of mouse CNR5; PCDH-alpha-13; Protocadherin alpha-13
Gene Aliases: CNR5; CNRN5; CNRS5; CRNR5; PCDH-ALPHA13; PCDHA13
UniProt ID: (Human) Q9Y5I0
Entrez Gene ID: (Human) 56136, (Rat) 116742
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support