Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
Promotions
View available promotion(s)
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
Immunogen sequence: MEIGWMHNRR QRQVLVFFVL LSLSGAGAEL GSYSVVEETE RGSFVANLGK DLGLGLTEMS TRKARIISQG NKQHLQLKAQ TGDLLINEKL DREELCGPTE PCILHFQVLM ENPLEIFQAE LRVIDINDHS PMFTEKEMIL KIPENSPLGT EFPLNHALDL DVGSNNVQNY KISPSSHFRV LIHEFRDGRK YPELVLDKEL DREEEPQLRL TLTALDGGSP PRSGTAQVRI EVVDINDNAP EFEQPIYKVQ IPENSPLGSL VATVSARDLD GGANGKISYT LFQPSEDISK TLEVNPMTGE VRLRKQVDFE MVTSYEVRIK ATDGGGLSGK CTLLLQVVDV NDNPPQVTMS ALTSPIAENS PEIVVAVFSV SDPDSGNNGK TISSIQEDLP FLLKPSVKNF YTLVTERALD REARAEYNIT LTVTDMGTPR LKTEHNITVQ ISDVNDNAPT FTQTSYTLFV RENNSPALHI GSVSATDRDS GTNAQVTYSL LPPQDPHLPL ASLVSINADN GHLFALRSLD YEALQAFEFR VGATDRGSPA LSREALVRVL VLDANDNSPL VLYPLQNGSA PCTELVPRAA EPGYLVTKVV AVDGDSGQNA WLSYQLLKAT EPGLFGVWAH NGEVRTARLL SERDAAKQRL VVLVKDNGEP PRSATATLHV LLVDGFSQPF LPLPEAAPGQ TQANSLTVYL VVALASVSSL FLFSVLLFVA VRLCRRSRAA SVGRCSMPEG PFPGRLVDVS GTGTLSQSYQ YEVCLTGGSE TSEFKFLKPI IPNFSP
This gene is a member of the protocadherin beta gene cluster, one of three related gene clusters tandemly linked on chromosome five. The gene clusters demonstrate an unusual genomic organization similar to that of B-cell and T-cell receptor gene clusters. The beta cluster contains 16 genes and 3 pseudogenes, each encoding 6 extracellular cadherin domains and a cytoplasmic tail that deviates from others in the cadherin superfamily. The extracellular domains interact in a homophilic manner to specify differential cell-cell connections. Unlike the alpha and gamma clusters, the transcripts from these genes are made up of only one large exon, not sharing common 3' exons as expected. These neural cadherin-like cell adhesion proteins are integral plasma membrane proteins. Their specific functions are unknown but they most likely play a critical role in the establishment and function of specific cell-cell neural connections.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: cadherin ME1; PCDH-beta-16; PCDHbeta 16; protocadherin beta 8a; Protocadherin beta-16; Protocadherin-3X
Gene Aliases: KIAA1621; ME1; PCDH-BETA16; PCDH3X; PCDHB16; PCDHB8a
UniProt ID: (Human) Q9NRJ7
Entrez Gene ID: (Human) 57717
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support