Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
NeoBiotechnologies
PCNA (polymerase delta auxiliary protein) is essential for DNA replication and is involved in DNA excision and mismatch repair pathways. PCNA binds to the CDK inhibitor p21, the structure-specific endonucleases Fen1 and XPG, and DNA cytosine 5-methyltransferase (MCMT). PCNA is a potentially useful marker of cells with proliferative potential and for identifying the proliferation status of tumor tissue (i.e. relevant to prognosis). PCNA is a marker for cells in early G1 phase and S phase of the cell cycle. PCNA is found in the nucleus and is a cofactor of DNA polymerase delta, and acts as a homotrimer and helps increase the processing of leading strand synthesis during DNA replication. In response to DNA damage, PCNA is ubiquitinated and is involved in the RAD6-dependent DNA repair pathway. Two transcript variants encoding the same protein have been found for PCNA. Pseudogenes of PCNA have been described on chromosome 4 and on the X chromosome.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CG9193-PA; CG9193-PB; Cyclin; DNA polymerase delta auxiliary protein; etID36690.10; HGCN8729; LD45889p; MGC8367; mutagen-sensitive 209; Mutagen-sensitive 209 protein; PCNA; PCNA-PA; PCNA-PB; Proliferating cell nuclear antigen; proliferating-cell nuclear antigen; Proliferating-cell-nuclear-antigen
Gene Aliases: 53/13; ATLD2; cb16; CG9193; Dmel\CG9193; Dmel_CG9193; DmPCNA; DmPCNA1; dPCNA; fa28e03; fb36g03; l(2)02448; l(2)56F; l(2)56Fa; l(2)s1534; mus 209; MUS209; PCNA; Pcna/cyclin; PCNA/mus209; PCNA1; PCNAR; POL30; wu:fa28e03; wu:fb36g03; YBR0811; YBR088C
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support