Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
Positive control: Rat brain tissue lysates, human stomach tissue.
Predicted band size: 74 kDa
Subcellular Location: Cell membrane; Perinuclear region.
The protein encoded by this gene is a member of the protein tyrosine phosphatase (PTP) family. PTPs are known to be signaling molecules that regulate a variety of cellular processes including cell growth, differentiation, mitotic cycle, and oncogenic transformation. This PTP possesses an extracellular region, a single transmembrane region, and a single intracellular catalytic domains, and thus represents a receptor-type PTP. The similar gene predominately expressed in mouse brain was found to associate with, and thus regulate the activity and cellular localization of MAP kinases. The rat counterpart of this gene was reported to be regulated by the nerve growth factor, which suggested the function of this gene in neuronal growth and differentiation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Ch-1 PTPase; Ch-1PTPase; ECPPT; NC-PTPCOM1; PC12-PTP1; protein tyrosine phosphatase Cr1PTPase; protein tyrosine phosphatase PCPTP1; protein tyrosine phosphatase, receptor type, R; protein-tyrosine phosphatase NC-PTPCOM1; Protein-tyrosine phosphatase PCPTP1; R-PTP-R; Receptor-type tyrosine-protein phosphatase R; Tyrosine phosphatase CBPTP
Gene Aliases: EC-PTP; ECPTP; PCPTP1; Ptp; PTP-SL; PTPBR7; PTPRQ; PTPRR
UniProt ID: (Human) Q15256, (Rat) O08617
Entrez Gene ID: (Human) 5801, (Rat) 94202
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support