Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
PA1-057 detects prohormone convertase 1 (PC1) from mouse pituitary extract.
PA1-057 has been successfully used in Western blot procedures. By Western blot, this antibody detects an ~80 kDa protein representing PC1 from mouse pituitary extract.
The PA1-057 immunogen is a synthetic peptide corresponding to residues S(111) V Q K D S A L D L F N D P M W N(127) of mouse PC1. This sequence is 88% and 82% conserved bovine and rat, respectively. The PA1-057 immunizing peptide (Cat. # PEP-164) is available for use in neutralization and control experiments.
The protein encoded by this gene belongs to the subtilisin-like proprotein convertase family. The members of this family are proprotein convertases that process latent precursor proteins into their biologically active products. This encoded protein is a type I proinsulin-processing enzyme that plays a key role in regulating insulin biosynthesis. It is also known to cleave proopiomelanocortin, prorenin, proenkephalin, prodynorphin, prosomatostatin and progastrin. Mutations in this gene are thought to cause obesity. This encoded protein is associated with carcinoid tumors. The use of alternate polyadenylation sites has been found for this gene. Multiple alternatively spliced transcript variants have been described for this gene but their full length nature is not known.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: EC 3.4.21.93; Furin homolog; NEC 1; Neuroendocrine convertase 1; PC1; PC3; Prohormone convertase 1; prohormone convertase 1/3; Propeptide-processing protease; Proprotein convertase 1
Gene Aliases: Att-1; Nec-1; Nec1; PC1; PC3; Pcsk1; Phpp-1; SPC3
UniProt ID: (Mouse) P63239
Entrez Gene ID: (Mouse) 18548
Molecular Function:
serine protease
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support