Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 93%
The protein encoded by this gene belongs to the subtilisin-like proprotein convertase family. The members of this family are proprotein convertases that process latent precursor proteins into their biologically active products. This encoded protein is a calcium-dependent serine endoprotease that can cleave precursor protein at their paired basic amino acid processing sites. Some of its substrates are - transforming growth factor beta related proteins, proalbumin, and von Willebrand factor. This gene is thought to play a role in tumor progression. Alternatively spliced transcript variants encoding different isoforms have been identified.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Paired basic amino acid cleaving enzyme 4; paired basic amino acid cleaving system 4; Proprotein convertase subtilisin/kexin type 6; SPC4; Subtilisin-like proprotein convertase 4; Subtilisin/kexin-like protease PACE4
Gene Aliases: PACE4; PCSK6; SPC4
UniProt ID: (Human) P29122
Entrez Gene ID: (Human) 5046
Molecular Function: serine protease
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support