Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 4
Positive Control: A431, HeLa, HepG2
Predicted Reactivity: Mouse (82%), Rat (83%), Rhesus Monkey (96%), Chimpanzee (100%)
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
PCSK9 is a proprotein convertase belonging to the proteinase K subfamily of the secretory subtilase family. This protein is synthesized as a soluble zymogen that undergoes autocatalytic intramolecular processing in the endoplasmic reticulum. The protein may function as a proprotein convertase. The protein plays a role in cholesterol homeostasis and may have a role in the differentiation of cortical neurons.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: convertase subtilisin/kexin type 9 preproprotein; hypercholesterolemia; NARC-1; neural apoptosis regulated convertase 1; Neural apoptosis-regulated convertase 1; OTTHUMP00000009833; PC9; Proprotein convertase 9; Proprotein convertase subtilisin/kexin type 9; Subtilisin/kexin-like protease PC9
Gene Aliases: FH3; HCHOLA3; LDLCQ1; NARC-1; NARC1; PC9; PCSK9; PSEC0052
UniProt ID: (Human) Q8NBP7
Entrez Gene ID: (Human) 255738
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support