Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Immunogen sequence: EFRTYSFPCY LPQPLINHAP RLDTDGIHLL SSLLLYESKS RMSAEAALSH SYFRSLGERV HQLEDTASIF SLKEIQLQKD PGYRGLAFQQ PGRGKNRRQS IF
May play a role in signal transduction cascades in terminally differentiated cells. [UniProt]
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Cell division protein kinase 18; Cyclin-dependent kinase 18; PCTAIRE protein kinase 3; PCTAIRE-motif protein kinase 3; Serine/threonine-protein kinase PCTAIRE-3
Gene Aliases: AA682070; CDK18; Ctk18; PCTAIRE; PCTAIRE3; PCTK3
UniProt ID: (Human) Q07002, (Mouse) Q04899
Entrez Gene ID: (Human) 5129, (Mouse) 18557
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support