Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
The recommended shelf life for this product is 1 year from date of receipt.
This gene belongs to the cytidylyltransferase family and is involved in the regulation of phosphatidylcholine biosynthesis. Mutations in this gene are associated with spondylometaphyseal dysplasia with cone-rod dystrophy. Alternatively spliced transcript variants have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CCT A; CCT-alpha; Choline-phosphate cytidylyltransferase A; CT A; CTP phosphocholine cytidylyl transferase; CTP:phosphocholine cytidylyltransferase A; CTP:phosphocholine cytidylyltransferase alpha; Phosphorylcholine transferase A
Gene Aliases: CCTA; Cctalpha; CT; CTA; CTalpha; CTPCT; Cttalpha; PCYT1; PCYT1A; SMDCRD
UniProt ID: (Human) P49585, (Mouse) P49586
Entrez Gene ID: (Human) 5130, (Mouse) 13026
Molecular Function:
transferase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support