Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Cell lysate).
Immunogen sequence: MTTLDDKLLG EKLQYYYSSS EDEDSDHEDK DRGRCAPASS SVPAEAELAG EGISVNTGPK GVINDWRRFK QLETEQREEQ CREMERLIKK LSMTCRSHLD EEEEQQKQKD LQEKISGKMT LKEFAIMNED QDDEEFLQQY RKQRMEEMRQ QLHKGPQFKQ VFEISSGEGF LDMIDKEQKS IVIMVHIYED GIPGTEAMNG CMICLAAEYP AVKFCKVKSS VIGASSQFTR NALPALLIYK GGELIGNFVR VTDQLGDDFF AVDLEAFLQE FGLLPEKEVL VLTSVRNSAT CHSEDSDLEI D
Phosducin-like protein is a putative modulator of heterotrimeric G proteins. The protein shares extensive amino acid sequence homology with phosducin, a phosphoprotein expressed in retina and pineal gland. Both phosducin-like protein and phosphoducin have been shown to regulate G-protein signaling by binding to the beta-gamma subunits of G proteins.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: PHLP; phosducin-like; Phosducin-like protein; RP11-465F21.2
Gene Aliases: PDCL; PHLOP1; PhLP; PhLP1
UniProt ID: (Human) Q13371
Entrez Gene ID: (Human) 5082
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support