Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 4
Phosducin-like proteins (PhLPs) are a conserved family of proteins with thioredoxin-like domains that were initially identified as modulators of G protein signaling. PDCL3 is highly homologous to PDCL and shares an N-terminal helix domain and a C-terminal thioredoxin-fold (Trx-fold) domain. Along with the related protein PDCL2, PDCL3 interacts with the chaperonin CCT and modulates CCT-mediated actin and tubulin folding. Modulation of PDCL3 levels by MAPK phosphorylation and RhoA-dependent changes also promote cytoskeletal remodeling.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: HTPHLP; IAP-associated factor VIAF1; phosducin-like 3; Phosducin-like protein 3; PhPL3; VIAF-1; Viral IAP-associated factor 1
Gene Aliases: 1110061A19Rik; C80025; HTPHLP; PDCL3; PHLP2A; PHLP3; VIAF; VIAF1
UniProt ID: (Human) Q9H2J4, (Mouse) Q8BVF2
Entrez Gene ID: (Human) 79031, (Mouse) 68833
Molecular Function: viral or transposable element protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support