Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Recommended positive controls: For western blot - Rat brain and pancreas.
Various cellular responses are regulated by the second messengers cAMP and cGMP. Phosphodiesterases, such as PDE10A, eliminate cAMP- and cGMP-mediated intracellular signaling by hydrolyzing the cyclic nucleotide to the corresponding nucleoside 5-prime monophosphate.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: cAMP and cAMP-inhibited cGMP 3',5'-cyclic phosphodiesterase 10A; dJ416F21.1 (phosphodiesterase 10A); dual-substrate cAMP and cGMP, cyclic nucleotide phosphodiesterase; FLJ11894; FLJ25677; PDE10A2 brain-specific isoform; phosphodiesterase 10A1 (PDE10A1); phosphodiesterase 10A7 (PDE10A7); testis-specific phosphodiesterase PDE10A2
Gene Aliases: ADSD2; HSPDE10A; IOLOD; LINC00473; PDE10A; PDE10A19; Pde10a3
UniProt ID: (Human) Q9Y233, (Mouse) Q8CA95, (Rat) Q9QYJ6
Entrez Gene ID: (Human) 10846, (Mouse) 23984, (Rat) 63885
Molecular Function:
phosphodiesterase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support