Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: KLYYREEAV AEDVLNVDAW QDGAVLQIGD VKYKVERNPP AFTELQLPRY IMAGFPVCPK LSLEFGDPAS SLFRWYKEAK PGAAEPEVGV PSSLSPSSPS SSWTETDVEE RVYTPSNADI GLRLKLHCTP GDGQRFGHSR ELESVCVVEA GPGTCTFDHR HLYTKKVTED ALIRTVSYNI LADTYAQTEF SRTVLYPYCA PYALELDYRQ NLIQKELTGY NADVICLQEV DRAVFSDSLV PALEAFGLEG VFRIKQHEGL ATFYRKSKFS LLSQHDISFY EALESDPLHK ELLEKLVLYP SAQEKVLQRS SVLQVSVLQS TKDSSKRICV ANTHLYWHPK GRFYLVSQVT (124-472 aa encoded by BC034978)
PDE12 is an enzyme that cleaves 2',5'-phosphodiester bond linking adenosines of the 5'-triphosphorylated oligoadenylates, triphosphorylated oligoadenylates referred as 2-5A modulates the 2-5A system. This enzyme degraded triphosphorylated 2-5A to produce AMP and ATP. It also cleaves 3',5'-phosphodiester bond of oligoadenylates and plays a role as a negative regulator of the 2-5A system that is one of the major pathways for antiviral and antitumor functions induced by interferons (IFNs). Suppression of this enzyme induces reduction of viral replication in Hela cells, thus counteracting the antiviral pathway probably by inhibiting the 2-5A system.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 2',5'-phosphodiesterase 12; 2'-PDE; 2'-phosphodiesterase; 2-PDE; Mitochondrial deadenylase
Gene Aliases: 2'-PDE; 2-PDE; E430028B21Rik; LRRGT00074; PDE12; RGD1310975
UniProt ID: (Human) Q6L8Q7, (Mouse) Q3TIU4
Entrez Gene ID: (Human) 201626, (Mouse) 211948, (Rat) 306231
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support