Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Promotions
View available promotion(s)
Promo Code: P5877545 Don't miss out: buy 3, only pay for 2 Extended to June 20: take advantage of our 3 for 2 promotion. Whether you are restocking essentials or trying something new, now is a great time to get more for your money!. Learn more
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
Peptide Sequence: LTPFPGFYPC SEIEDPAEKG DRKLNKGLNR NSLPTPQLRR SSGTSGLLPV EQSSRWDRNN GKRPHQEFGI SSQGCYLNGP FNSNLLTIPK QRSSSVSLTH
Cyclic nucleotide phosphodiesterase with a dual-specificity for the second messengers cAMP and cGMP, which are key regulators of many important physiological processes. May play a role in fat metabolism. Regulates cAMP binding of RAPGEF3. Through simultaneous binding to RAPGEF3 and PIK3R6 assembles a signaling complex in which the PI3K gamma complex is activated by RAPGEF3 and which is involved in angiogenesis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CGI-PDE B; CGIP1; CGIPDE1; cGMP-inhibited 3',5'-cyclic phosphodiesterase 3B; Cyclic GMP-inhibited phosphodiesterase B; cyclic nucleotide phosphodiesterase; phosphodiesterase 3B, cGMP-inhibited
Gene Aliases: cGIPDE1; HcGIP1; PDE3B
UniProt ID: (Human) Q13370
Entrez Gene ID: (Human) 5140
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support