Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
PDE6C is the alpha-prime subunit of cone phosphodiesterase, which is composed of a homodimer of two alpha-prime subunits and 3 smaller proteins of 11, 13, and 15 kDa.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: cGMP phosphodiesterase 6C; cGMP phosphodiesterase alpha subunit (PDE) (EC 3.1.4.17); Cone cGMP-specific 3',5'-cyclic phosphodiesterase subunit alpha'; cone photoreceptor cyclic nucleotide phosphodiesterase alpha'-subunit; PDE V-C1; phosphodiesterase 6C, cGMP-specific, cone, alpha prime; phosphodiesterase, cyclic GMP (cone receptor)
Gene Aliases: ACHM5; COD4; cpfl1; PDE6C; PDEA2
UniProt ID: (Bovine) P16586, (Human) P51160, (Mouse) Q91ZQ1
Entrez Gene ID: (Bovine) 281975, (Human) 5146, (Rat) 361752, (Mouse) 110855
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support