Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
PA1-31148 detects PDE9A from rat, human, and mouse samples.
The protein encoded by this gene catalyzes the hydrolysis of cAMP and cGMP to their corresponding monophosphates. The encoded protein plays a role in signal transduction by regulating the intracellular concentration of these cyclic nucleotides. Multiple transcript variants encoding several different isoforms have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CGMP-specific 3',5'-cyclic phosphodiesterase type 9; FLJ90181; High affinity cGMP-specific 3',5'-cyclic phosphodiesterase 9A; phosphodiesterase PDE9A21
Gene Aliases: HSPDE9A2; Pde8b; PDE9A; PDE9A1
UniProt ID: (Human) O76083, (Rat) Q8QZV1, (Mouse) O70628
Entrez Gene ID: (Human) 5152, (Rat) 191569, (Mouse) 18585
Molecular Function:
phosphodiesterase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support