Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 4
Anti-PDH Western Blot Kit; Western Blot kit (Cocktail of monoclonal antibodies) specific to Human, bovine PDH. This antibody is validated for use in Western Blot.
C-V-OSCP (clone ) 4C11C10D12 - Antigen Human rec. mature OSCP
Amount: 12.5 µg , Working concentration: 0.5 µg/mL
PDH-E1-alpha (clone) 9H9AF5 - Human rec. PDH-E1-a
Amount: 25 µg , Working concentration: 1 µg/mL
PDH-E1-beta (clone) 17A5E2H8 - Porcine PDH
Amount: 62.5 µg, Working concentration: 2.5 µg/mL
PDH-E2 (clone) 15D3G9C11 - Porcine PDH
Amount: 25 µg, Working concentration: 1 µg/mL
PDH-E2/E3bp (clone) 13G2AE2BH5 - Porcine PDH
Amount: 25 µg, Working concentration: 1 µg/mL
Pyruvate dehydrogenase (PDH) is a mitochondrial multienzyme complex that catalyzes the oxidative decarboxylation of pyruvate and is one of the major enzymes responsible for the regulation of homeostasis of carbohydrate fuels in mammals. The enzymatic activity is regulated by a phosphorylation/dephosphorylation cycle. Phosphorylation of PDH by a specific pyruvate dehydrogenase kinase (PDK) results in inactivation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 13G2AE2BH5; 15D3G9C11; 17A5E2H8; 4C11C10D12; 9H9AF5; PDHE1-A type I; PDHE1-A type II; PDHE1-B; pyruvate dehydrogenase; pyruvate dehydrogenase (lipoamide) alpha 1; pyruvate dehydrogenase (lipoamide) alpha 2; pyruvate dehydrogenase (lipoamide) beta; pyruvate dehydrogenase alpha 1; pyruvate dehydrogenase alpha 2; pyruvate dehydrogenase complex, E1-alpha polypeptide 1; pyruvate dehydrogenase E1 alpha 1; pyruvate dehydrogenase E1 alpha 1 subunit; pyruvate dehydrogenase E1 alpha 2; pyruvate dehydrogenase E1 alpha 2 subunit; pyruvate dehydrogenase E1 beta subunit; pyruvate dehydrogenase E1 component alpha subunit; PYRUVATE DEHYDROGENASE E1 COMPONENT ALPHA SUBUNIT, TESTIS-SPECIFIC FORM, MITOCHONDRIAL PRECURSOR (PDHE1-A TYPE II); Pyruvate dehydrogenase E1 component subunit alpha, somatic form, mitochondrial; Pyruvate dehydrogenase E1 component subunit alpha, testis-specific form, mitochondrial; Pyruvate dehydrogenase E1 component subunit beta, mitochondrial; pyruvate dehydrogenase E1 subunit; pyruvate dehydrogenase E1alpha subunit; pyruvate dehydrogenase, E1 beta polypeptide; pyruvate dehydrogenase, E1-alpha polypeptide, testis specific; RP11-723P2.1
Gene Aliases: 2610103L06Rik; AL024199; C81408; PDHA; Pdha-1; Pdha-2; PDHA1; PDHA2; PDHAD; PDHAL; PDHB; PDHBD; PDHCE1A; PDHE1-B; PHE1A; PHE1B
UniProt ID: (Human) P08559, (Bovine) A7MB35, (Human) P29803, (Human) P11177, (Bovine) P11966, (Mouse) P35486, (Mouse) P35487, (Rat) Q06437, (Rat) P49432, (Mouse) Q9D051
Entrez Gene ID: (Human) 5160, (Bovine) 407109, (Bovine) 768012, (Human) 5161, (Human) 5162, (Bovine) 613610, (Rat) 29554, (Mouse) 18597, (Mouse) 18598, (Rat) 117098, (Rat) 289950, (Mouse) 68263
Molecular Function: dehydrogenase oxidoreductase metabolite interconversion enzyme
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support