Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 4
Immunogen sequence: GVECEVINMR TIRPMDMETI EASVMKTNHL VTVEGGWPQF GVGAEICARI MEGPAFNFLD APAVRVTGAD VPMPYAKILE DNSIPQVKDI IFAIKKTLNI
PDHB (Pyruvate dehydrogenase E1 component subunit beta, mitochondrial) is also named as PHE1B. The enzyme, which is found in mitochondria, is one of the component enzymes of the pyruvate dehydrogenase multienzyme complex (PDH). It catalyzes the first reaction of an oxidative decarboxylation sequence converting pyruvate to acetyl-CoA and CO(2). Defects in PDHB are the cause of pyruvate dehydrogenase E1-beta deficiency (PDHBD).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: PDHE1-B; Pyruvate dehydrogenase E1 component subunit beta, mitochondrial; pyruvate dehydrogenase, E1 beta polypeptide
Gene Aliases: 2610103L06Rik; AL024199; C81408; PDHB; PDHBD; PDHE1-B; PHE1B
UniProt ID: (Human) P11177, (Rat) P49432, (Mouse) Q9D051
Entrez Gene ID: (Human) 5162, (Rat) 289950, (Mouse) 68263
Molecular Function: dehydrogenase oxidoreductase metabolite interconversion enzyme
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support