Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 6
Positive Control: Rat heart tissue lysate, rat brain tissue, human colon cancer tissue, human fetal skeletal muscle tissue, mouse cerebellum tissue, SH-SY-5Y.
PDK2 inhibits the mitochondrial pyruvate dehydrogenase complex by phosphorylation of the E1 alpha subunit, thus contributing to the regulation of glucose metabolism.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: [Pyruvate dehydrogenase (acetyl-transferring)] kinase isozyme 2, mitochondrial; PDH kinase 2; pyruvate dehydrogenase; Pyruvate dehydrogenase kinase isoform 2; pyruvate dehydrogenase kinase, isoenzyme 2; pyruvate dehydrogenase, lipoamide, kinase isozyme 2, mitochondrial
Gene Aliases: PDHK2; PDK2; PDKII
UniProt ID: (Human) Q15119
Entrez Gene ID: (Human) 5164
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support