Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: GVIEYKEKF GFDPFISTNI QYFLDRFYTN RISFRMLINQ HTLLFGGDTN PVHPKHIGSI DPTCNVADVV KDAYETAKML CEQYYLVAPE LEVEEFNAKA PDKPIQVVYV PSHLFHMLFE LFKNSMRATV ELYEDRKEGY PAVKTLVTLG KEDLSIKISD LGGGVPLRKI DRLFNYMYST APRPSLEPTR AAPLAGFGYG LPISRLYARY FQGDLKLYSM EGVGTDAVIY LKALSSESFE RL (129-369 aa encoded by BC015948)
PDK3 inhibits the mitochondrial pyruvate dehydrogenase complex by phosphorylation of the E1 alpha subunit, thus contributing to the regulation of glucose metabolism.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: [Pyruvate dehydrogenase (acetyl-transferring)] kinase isozyme 3, mitochondrial; [Pyruvate dehydrogenase [lipoamide]] kinase isozyme 3, mitochondrial; pyruvate dehydrogenase; pyruvate dehydrogenase kinase 3; Pyruvate dehydrogenase kinase isoform 3; pyruvate dehydrogenase kinase, isoenzyme 3; pyruvate dehydrogenase kinase, isozyme 3; pyruvate dehydrogenase, lipoamide, kinase isozyme 3, mitochondrial
Gene Aliases: 2610001M10Rik; AI035637; CMTX6; GS1-358P8.4; PDHK3; PDK3
UniProt ID: (Human) Q15120, (Mouse) Q922H2
Entrez Gene ID: (Human) 5165, (Mouse) 236900
Molecular Function:
non-receptor serine/threonine protein kinase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support