Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Recommended positive controls: HepG2, mouse liver, PDK3-transfected 293T.
Predicted reactivity: Mouse (96%), Rat (96%), Chicken (93%), Bovine (98%).
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
PDK3 inhibits the mitochondrial pyruvate dehydrogenase complex by phosphorylation of the E1 alpha subunit, thus contributing to the regulation of glucose metabolism.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: [Pyruvate dehydrogenase (acetyl-transferring)] kinase isozyme 3, mitochondrial; [Pyruvate dehydrogenase [lipoamide]] kinase isozyme 3, mitochondrial; pyruvate dehydrogenase; pyruvate dehydrogenase kinase 3; Pyruvate dehydrogenase kinase isoform 3; pyruvate dehydrogenase kinase, isoenzyme 3; pyruvate dehydrogenase kinase, isozyme 3; pyruvate dehydrogenase, lipoamide, kinase isozyme 3, mitochondrial
Gene Aliases: 2610001M10Rik; AI035637; CMTX6; GS1-358P8.4; PDHK3; PDK3
UniProt ID: (Human) Q15120, (Mouse) Q922H2
Entrez Gene ID: (Human) 5165, (Mouse) 236900
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support