Invitrogen antibodies—maximize lab budgets with advanced validation & coverage

Shop antibodies

Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Primary Antibodies
  • Search All
  • Popular Product Areas
  • TaqMan Assays
  • Primary Antibodies
  • Secondary Antibodies
  • Isotype Controls
  • Proteins & Peptides
  • ELISA Kits
  • All Documents & Support
  • Certificates
  • SDS
  • Manuals & Protocols
  • Product FAQs
    Search button Close
            • Order Status
            • Quick Order
            • Sign in
              Sign in
              Don't have an account ? Create Account
              • Account
              • Check Order Status
              • Aspire Member Program
              • Connect: Lab, Data, Apps
              • Custom Products & Projects
              • Services Central

            Disclaimer

            Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

            Loading ...
            • Primary Antibodies ›
            • PDK4 Antibodies

            Proteintech

            PDK4 Polyclonal Antibody

            47 References
            View all (12) PDK4 antibodies
            Datasheet
            Protocols
            Questions & Answers
            Datasheet
            Protocols
            Questions & Answers

            Cite PDK4 Polyclonal Antibody

            • Antibody Testing Data (15)
            Application
            PDK4 Antibody in Immunocytochemistry (ICC/IF)
            Group 53 Created with Sketch.

            FIGURE: 1 / 15

            PDK4 Antibody (12949-1-AP) in ICC/IF

            Immunofluorescent analysis of ( 4% PFA) fixed HepG2 cells using 12949-1-AP (PDK4 antibody) at dilution of 1:50 and Alexa Fluor 488-conjugated AffiniPure Goat Anti-Rabbit IGG (H+L).
            View Product

            PDK4 Antibody in Immunocytochemistry (ICC/IF)
            PDK4 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
            PDK4 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
            PDK4 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
            PDK4 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
            PDK4 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
            PDK4 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
            PDK4 Antibody in Western Blot (WB)
            PDK4 Antibody in Western Blot (WB)
            PDK4 Antibody in Western Blot (WB)
            PDK4 Antibody in Western Blot (WB)
            PDK4 Antibody in Western Blot (WB)
            PDK4 Antibody in Western Blot (WB)
            PDK4 Antibody in Western Blot (WB)
            PDK4 Antibody in Western Blot (WB)

            Product Details

            12949-1-AP

            Applications
            Tested Dilution
            Publications

            Western Blot (WB)

            1:1,000-1:3,000
            View 39 publications 39 publications

            Immunohistochemistry (IHC)

            -
            View 5 publications 5 publications

            Immunohistochemistry (Paraffin) (IHC (P))

            1:50-1:500
            -

            Immunocytochemistry (ICC/IF)

            1:50-1:500
            View 3 publications 3 publications
            Product Specifications

            Species Reactivity

            Human, Mouse, Rat

            Published species

            Human, Mouse, Rat

            Host/Isotype

            Rabbit / IgG

            Class

            Polyclonal

            Type

            Antibody

            Immunogen

            PDK4 Fusion Protein Ag3629 (61-412 aa encoded by BC040239)
            View immunogen

            Conjugate

            Unconjugated

            Form

            Liquid

            Concentration

            0.19 mg/mL

            Purification

            Antigen affinity chromatography

            Storage buffer

            PBS, pH 7.3, with 50% glycerol

            Contains

            0.02% sodium azide

            Storage conditions

            -20°C

            Shipping conditions

            Wet ice (domestic); Dry ice (international)

            Product Specific Information

            Immunogen sequence: LPVRLANIL KEIDILPTQL VNTSSVQLVK SWYIQSLMDL VEFHEKSPDD QKALSDFVDT LIKVRNRHHN VVPTMAQGII EYKDACTVDP VTNQNLQYFL DRFYMNRIST RMLMNQHILI FSDSQTGNPS HIGSIDPNCD VVAVVQDAFE CSRMLCDQYY LSSPELKLTQ VNGKFPDQPI HIVYVPSHLH HMLFELFKNA MRATVEHQEN QPSLTPIEVI VVLGKEDLTI KISDRGGGVP LRIIDRLFSY TYSTAPTPVM DNSRNAPLAG FGYGLPISRL YAKYFQGDLN LYSLSGYGTD AIIYLKALSS ESIEKLPVFN KSAFKHYQMS SEADDWCIPS REPKNLAKEV AM (61-412 aa encoded by BC040239)

            Target Information

            PDK4 (PDHK4), along with the other PDHKs, regulates glucose metabolism by phosphorylating the E1 alpha subunit of the mitochondrial pyruvate dehydrogenase complex. Inhibition of PDHKs is viewed as a potential therapeutic treatment for type II diabetes. This protein is located in the matrix of the mitrochondria and it's expression is regulated by glucocorticoids, retinoic acid and insulin. PDK4 inhibits the mitochondrial pyruvate dehydrogenase complex by phosphorylation of the E1 alpha subunit, thus contributing to the regulation of glucose metabolism.

            For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

            Bioinformatics

            Protein Aliases: [Pyruvate dehydrogenase (acetyl-transferring)] kinase isozyme 4, mitochondrial; [Pyruvate dehydrogenase [lipoamide]] kinase isozyme 4, mitochondrial; kinase isozyme 4; lipoamide; pyruvate dehydrogenase; pyruvate dehydrogenase kinase 4; Pyruvate dehydrogenase kinase isoform 4; pyruvate dehydrogenase kinase, isoenzyme 4; pyruvate dehydrogenase kinase, isozyme 4; pyruvate dehydrogenase, lipoamide, kinase isozyme 4, mitochondrial; pyruvate dehydrogenate kinase 4

            View more View less

            Gene Aliases: AV005916; PDHK4; PDK4

            View more View less

            UniProt ID: (Human) Q16654, (Rat) O54937, (Mouse) O70571

            View more View less

            Entrez Gene ID: (Human) 5166, (Rat) 89813, (Mouse) 27273

            View more View less

            Function(s)

            protein kinase activity pyruvate dehydrogenase (acetyl-transferring) kinase activity ATP binding nucleotide binding protein serine/threonine/tyrosine kinase activity kinase activity transferase activity non-receptor serine/threonine protein kinase

            Process(es)

            positive regulation of defense response to virus by host glucose metabolic process protein phosphorylation regulation of pH insulin receptor signaling pathway cellular response to starvation regulation of acetyl-CoA biosynthetic process from pyruvate regulation of cellular ketone metabolic process regulation of glucose metabolic process regulation of fatty acid biosynthetic process glucose homeostasis response to starvation regulation of bone resorption regulation of fatty acid oxidation cellular response to fatty acid reactive oxygen species metabolic process xenophagy negative regulation of anoikis acetyl-CoA biosynthetic process from pyruvate carbohydrate metabolic process phosphorylation
            It has to be done as per old AB suggested Products section.
            guarantee_icon

            Performance Guarantee

            If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

            Learn more
            help_icon

            We're here to help

            Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

            Contact tech support
            Ordering Plus Icon Minus Icon
            • Order Status
            • Order Help
            • Quick Order
            • Supply Center
            • eProcurement
            Support Plus Icon Minus Icon
            • Help and Support
            • Contact Us
            • Technical Support Centers
            • Documents and Certificates
            • Report a Site Issue
            Resources Plus Icon Minus Icon
            • Learning Centers
            • Promotions
            • Events and Webinars
            • Social Media
            About Thermo Fisher Plus Icon Minus Icon
            • About Us
            • Careers
            • Investors
            • News
            • Social Responsibility
            • Trademarks
            • Consumer Health Data Privacy Policy
            Our Portfolio Plus Icon Minus Icon
            • Thermo Scientific
            • Applied Biosystems
            • Invitrogen
            • Gibco
            • Ion Torrent
            • Fisher Scientific
            • Unity Lab Services
            • Patheon
            • PPD
            • Terms & Conditions
            • Privacy Information Center
            • Price & Freight Policy
            • Cookie Preferences

              Your choices regarding cookies on this site

              We and our affiliates and vendors use cookies and similar technologies to operate our sites, recognize visitors to our sites, provide secure log-in, collect statistics to optimize site functionality, and deliver content tailored to your interests. Click Accept All to accept all cookies and go directly to the site, click Reject All to reject all but cookies strictly necessary to the functioning of this site (required cookies), or click on Manage Settings to see detailed descriptions of the types of cookies and choose whether to accept certain cookies while on the site.
            © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
            United States flag icon
            United States

            Your items have has been added!


            Host server : magellan-search-green-56756dfb8b-z2vcc:80/100.66.79.199:80.
            git-commit: de6905f249d25e22729e613aef254bb544bd8b7c
            git-url: https://github.com/thermofisher/magellan-search
            git-branch: release/2.31.1-2025.06.25-1.0