Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Primary Antibodies
  • Search All
  • Popular Product Areas
  • TaqMan Assays
  • Primary Antibodies
  • Secondary Antibodies
  • Isotype Controls
  • Proteins & Peptides
  • ELISA Kits
  • All Documents & Support
  • Certificates
  • SDS
  • Manuals & Protocols
  • Product FAQs
    Search button Close
            Target of Interest
            Application
            Target Species
            • Order Status
            • Quick Order
            • Sign in
              Sign in
              Don't have an account ? Create Account
              • Account
              • Check Order Status
              • Aspire Member Program
              • Connect: Lab, Data, Apps
              • Custom Products & Projects
              • Services Central

            Disclaimer

            Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

            Loading ...
            • Primary Antibodies ›
            • PDLIM7 Antibodies

            Proteintech

            PDLIM7 Polyclonal Antibody

            15 References
            View all (16) PDLIM7 antibodies
            Datasheet
            Protocols
            Questions & Answers
            Datasheet
            Protocols
            Questions & Answers

            Cite PDLIM7 Polyclonal Antibody

            • Antibody Testing Data (8)
            Application
            PDLIM7 Antibody in Immunocytochemistry (ICC/IF)
            Group 53 Created with Sketch.

            FIGURE: 1 / 8

            PDLIM7 Antibody (10221-1-AP) in ICC/IF

            Immunofluorescent analysis of ( -20°C Ethanol ) fixed SKOV-3 cells using 10221-1-AP (PDLIM7 antibody) at dilution of 1:50 and Alexa Fluor 488-conjugated AffiniPure Goat Anti-Rabbit IGG (H+L).
            View Product

            PDLIM7 Antibody in Immunocytochemistry (ICC/IF)
            PDLIM7 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
            PDLIM7 Antibody in Western Blot (WB)
            PDLIM7 Antibody in Western Blot (WB)
            PDLIM7 Antibody in Western Blot (WB)
            PDLIM7 Antibody in Western Blot (WB)
            PDLIM7 Antibody in Immunoprecipitation (IP)
            PDLIM7 Antibody in Immunoprecipitation (IP)

            Product Details

            10221-1-AP

            Applications
            Tested Dilution
            Publications

            Western Blot (WB)

            1:1,000-1:4,000
            View 9 publications 9 publications

            Immunohistochemistry (IHC)

            -
            View 4 publications 4 publications

            Immunohistochemistry (Paraffin) (IHC (P))

            1:20-1:200
            -

            Immunocytochemistry (ICC/IF)

            1:20-1:200
            View 1 publication 1 publication

            Immunoprecipitation (IP)

            0.5-4.0 µg
            View 1 publication 1 publication
            Product Specifications

            Species Reactivity

            Human

            Published species

            Human, Mouse

            Host/Isotype

            Rabbit / IgG

            Class

            Polyclonal

            Type

            Antibody

            Immunogen

            PDLIM7 Fusion Protein Ag0264 (3-289 aa encoded by BC001093)
            View immunogen

            Conjugate

            Unconjugated

            Form

            Liquid

            Concentration

            0.4 mg/mL

            Purification

            Antigen affinity chromatography

            Storage buffer

            PBS, pH 7.3, with 50% glycerol

            Contains

            0.02% sodium azide

            Storage conditions

            -20°C

            Shipping conditions

            Wet ice (domestic); Dry ice (international)

            Product Specific Information

            Immunogen sequence: SFKVVLEGP APWGFRLQGG KDFNVPLSIS RLTPGGKAAQ AGVAVGDWVL SIDGENAGSL THIEAQNKIR ACGERLSLGL SRAQPVQSKP QKASAPAADP PRYTFAPSVS LNKTARPFGA PPPADSAPQQ NGQPLRPLVP DASKQRLMEN TEDWRPRPGT GQSRSFRILA HLTGTEFMQD PDEEHLKKSS QVPRTEAPAP ASSTPQEPWP GPTAPSPTSR PPWAVDPAFA ERYAPDKTST VLTRHSQPAT PTPLQSRTSI VQAAAGGVPG GGSNNGKTPV CHQCHKVI (3-289 aa encoded by BC001093)

            Target Information

            The protein encoded by this gene is representative of a family of proteins composed of conserved PDZ and LIM domains. LIM domains are proposed to function in protein-protein recognition in a variety of contexts including gene transcription and development and in cytoskeletal interaction. The LIM domains of this protein bind to protein kinases, whereas the PDZ domain binds to actin filaments. The gene product is involved in the assembly of an actin filament-associated complex essential for transmission of ret/ptc2 mitogenic signaling. The biological function is likely to be that of an adapter, with the PDZ domain localizing the LIM-binding proteins to actin filaments of both skeletal muscle and nonmuscle tissues. Alternative splicing of this gene results in multiple transcript variants.

            For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

            Bioinformatics

            Protein Aliases: 1110003B01Rik; LIM domain protein; LIM mineralization protein; Lim mineralization protein 3; LMP; PDZ and LIM domain 7 (enigma); PDZ and LIM domain protein 7; Protein enigma

            View more View less

            Gene Aliases: ENIGMA; LMP1; LMP3; PDLIM7

            View more View less

            UniProt ID: (Human) Q9NR12

            View more View less

            Entrez Gene ID: (Human) 9260

            View more View less

            Function(s)

            protein binding zinc ion binding actin or actin-binding cytoskeletal protein

            Process(es)

            ossification receptor-mediated endocytosis multicellular organism development actin cytoskeleton organization cell differentiation positive regulation of osteoblast differentiation
            It has to be done as per old AB suggested Products section.
            guarantee_icon

            Performance Guarantee

            If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

            Learn more
            help_icon

            We're here to help

            Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

            Contact tech support
            Ordering Plus Icon Minus Icon
            • Order Status
            • Order Help
            • Quick Order
            • Supply Center
            • eProcurement
            Support Plus Icon Minus Icon
            • Help and Support
            • Contact Us
            • Technical Support Centers
            • Documents and Certificates
            • Report a Site Issue
            Resources Plus Icon Minus Icon
            • Learning Centers
            • Promotions
            • Events and Webinars
            • Social Media
            About Thermo Fisher Plus Icon Minus Icon
            • About Us
            • Careers
            • Investors
            • News
            • Social Responsibility
            • Trademarks
            • Consumer Health Data Privacy Policy
            Our Portfolio Plus Icon Minus Icon
            • Thermo Scientific
            • Applied Biosystems
            • Invitrogen
            • Gibco
            • Ion Torrent
            • Fisher Scientific
            • Unity Lab Services
            • Patheon
            • PPD
            • Terms & Conditions
            • Privacy Information Center
            • Price & Freight Policy
            • Cookie Preferences

              Your choices regarding cookies on this site

              We and our affiliates and vendors use cookies and similar technologies to operate our sites, recognize visitors to our sites, provide secure log-in, collect statistics to optimize site functionality, and deliver content tailored to your interests. Click Accept All to accept all cookies and go directly to the site, click Reject All to reject all but cookies strictly necessary to the functioning of this site (required cookies), or click on Manage Settings to see detailed descriptions of the types of cookies and choose whether to accept certain cookies while on the site.
            © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
            United States flag icon
            United States

            Your items have has been added!


            Host server : magellan-search-green-56756dfb8b-wbd5c:80/100.66.79.199:80.
            git-commit: de6905f249d25e22729e613aef254bb544bd8b7c
            git-url: https://github.com/thermofisher/magellan-search
            git-branch: release/2.31.1-2025.06.25-1.0