Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
Decreases the sensitivity of PDP1 to magnesium ions, and this inhibition is reversed by the polyamine spermine.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: PDPr; Pyruvate dehydrogenase phosphatase regulatory subunit, mitochondrial
Gene Aliases: 4930402E16Rik; AU018056; KIAA1990; mKIAA1990; PDP3; PDPR
UniProt ID: (Human) Q8NCN5, (Mouse) Q7TSQ8
Entrez Gene ID: (Human) 55066, (Mouse) 319518
Molecular Function: dehydrogenase methyltransferase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support