Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
In direct ELISAs, approximately 5% cross-reactivity with recombinant human PDZD2 and recombinant mouse PDZD2 is observed.
Reconstitute at 0.2 mg/mL in sterile PBS.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ATPase-interacting PDZ protein; PDZ domain-containing protein 11; Plasma membrane calcium ATPase-interacting single-PDZ protein; PMCA-interacting single-PDZ protein
Gene Aliases: AIPP1; HSPC227; PDZD11; PDZK11; PISP; UNQ6486/PRO21335
UniProt ID: (Human) Q5EBL8
Entrez Gene ID: (Human) 51248
Molecular Function: cell junction protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support