Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Sandwich ELISA (Recombinant protein).
Immunogen sequence: RQVSKTRLSE PGTDLVEPSP KHTPNTSDNE GSDTEVCGPN SPSKRGNSTG IKLVRKEGGL DDSVFIAVKE IGRDLYRGLP TEERIQKLEF MLDKLQNEID QELEHNNSLV
PDZD8 plays a role in the regulation of cell morphology and cytoskeletal organization. It may inhibit herpes simplex virus 1 infection at an early stage and may promote retroviral infection.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: bA129M16.2; FLJ34427; PDZ domain-containing protein 8; Sarcoma antigen NY-SAR-84/NY-SAR-104
Gene Aliases: PDZD8; PDZK8
UniProt ID: (Human) Q8NEN9
Entrez Gene ID: (Human) 118987
Molecular Function: scaffold/adaptor protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support