Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
PEA15 is a death effector domain (DED)-containing protein predominantly expressed in the central nervous system, particularly in astrocytes.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 15 kDa phosphoprotein enriched in astrocytes; Astrocytic phosphoprotein PEA-15; homolog of mouse MAT-1 oncogene; mammary transforming gene 1; mammary transforming gene 1, mouse, homolog of; OTTHUMP00000029887; PED; phosphoprotein enriched in astrocytes 15; phosphoprotein enriched in astrocytes 15A; Phosphoprotein enriched in diabetes
Gene Aliases: HMAT1; HUMMAT1H; MAT1; MAT1H; PEA-15; PEA15; Pea15a; PED; Pkcs15
UniProt ID: (Human) O00511, (Rat) Q5U318, (Mouse) Q62048
Entrez Gene ID: (Human) 8682, (Bovine) 510586, (Rat) 364052, (Mouse) 18611
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support