Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Sandwich ELISA (Recombinant protein).
Immunogen sequence: TPPPLPKKMI IRANTEPISK DLQKSMESSL CVMANPTYDI DPNWDASSAG SSISYELKGL DIESYDSLER PLRKERPVPS AANSISSLTT LSIKDRFSNS
Tyrosine kinase that may play a role in cell spreading and migration on fibronectin. May directly or indirectly affect phosphorylation levels of cytoskeleton-associated proteins MAPK1/ERK and PXN.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Inactive tyrosine-protein kinase PEAK1; NKF3 kinase family member; Pseudopodium-enriched atypical kinase 1; Sugen kinase 269; Tyrosine-protein kinase SgK269
Gene Aliases: KIAA2002; PEAK1; SGK269
UniProt ID: (Human) Q9H792
Entrez Gene ID: (Human) 79834
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support