Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MASWAKGRS YLAPGLLQGQ VAIVTGGATG IGKAIVKELL ELGSNVVIAS RKLERLKSAA DELQANLPPT KQARVIPIQC NIRNEEEVNN LVKSTLDTFG KINFLVNNGG GQFLSPAEHI SSKGWHAVLE TNLTGTFYMC KAVYSSWMKE HGGSIVNIIV PTKAGFPLAV HSGAARAGVY NLTKSLALEW ACSGIRINCV APGVIYSQTA VENYGSWGQS FFEGSFQKIP AKRIGVPEEV SSVVCFLLSP AASFITGQSV DVDGGRSLYT HSYEVPDHDN WPKGAGDLSV VKKMKETFKE KAKL (1-303 aa encoded by BC002529)
PECR (Peroxisomal Trans-2-Enoyl-CoA Reductase) is a Protein Coding gene. Among its related pathways are Metabolism and Allograft rejection. Gene Ontology (GO) annotations related to this gene include signaling receptor binding and coenzyme binding. An important paralog of this gene is DECR1.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 2,4-dienoyl-CoA reductase-related protein; DCR-RP; HPDHase; perosisomal 2-enoyl-CoA reductase; Peroxisomal trans-2-enoyl-CoA reductase; putative short chain alcohol dehydrogenase; pVI-ARL; Short chain dehydrogenase/reductase family 29C member 1; short chain dehydrogenase/reductase family 29C, member 1; TERP
Gene Aliases: 2400003B18Rik; DCRRP; HPDHASE; HSA250303; PECR; PRO1004; PVIARL; SDR29C1; TERP
UniProt ID: (Human) Q9BY49, (Mouse) Q99MZ7
Entrez Gene ID: (Human) 55825, (Mouse) 111175
Molecular Function:
oxidoreductase
reductase
metabolite interconversion enzyme
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support