Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Promotions
View available promotion(s)
Promo Code: P5906365 Don't miss out: buy 3, only pay for 2 Extended to June 27: take advantage of our 3 for 2 promotion. Whether you are restocking essentials or trying something new, now is a great time to get more for your money!. Learn more
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
FIGURE: 1 / 6
Recommended positive controls: 293T, HeLa, HepG2, PC-12, Rat2, , Neuro 2A, C8D30, NIH-3T3, Raw264.7, C2C12.
Predicted reactivity: Human (99%), Mouse (97%), Rat (97%), Zebrafish (82%), Xenopus laevis (83%), Chicken (95%), Rhesus Monkey (100%), Bovine (98%).
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
PELO encodes a protein which contains a conserved nuclear localization signal. The encoded protein may have a role in spermatogenesis, cell cycle control, and in meiotic cell division.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: hPelota; Protein Dom34 homolog; Protein pelota homolog
Gene Aliases: AA409897; AI852218; CGI-17; PELO; PRO1770
UniProt ID: (Human) Q9BRX2, (Mouse) Q80X73, (Rat) Q5XIP1
Entrez Gene ID: (Human) 53918, (Mouse) 105083, (Rat) 294754
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support