Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MKRSGNPGAE VTNSSVAGPD CCGGLGNIDF RQADFCVMTR LLGYVDPLDP SFVAAVITIT FNPLYWNVVA RWEHKTRKLS RAFGSPYLAC YSLSVTILLL NFLRSHCFTQ AMLSQPRMES LDTPAAYSLG LALLGLGVVL VLSSFFALGF AGTFLGDYFG ILKEARVTVF PFNILDNPMY WGSTANYLGW AIMHASPTGL LLTVLVALTY IVALLYEEPF TAEIYRQKAS GSHKRS
This gene encodes an enzyme which converts phosphatidylethanolamine to phosphatidylcholine by sequential methylation in the liver. The protein localizes to the endoplasmic reticulum and mitochondria-associated membranes. The gene is within the Smith-Magenis syndrome region on chromosome 17. Alternate splicing of this gene results in three transcript variants encoding two different isoforms.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: MGC2483; PEAMT; PEMT2; Phosphatidylethanolamine N-methyltransferase; Phospholipid methyltransferase; PLMT
Gene Aliases: PEAMT; PEMPT; PEMT; PEMT2; PNMT
UniProt ID: (Human) Q9UBM1
Entrez Gene ID: (Human) 10400
Molecular Function: methyltransferase transferase metabolite interconversion enzyme
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support