Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:32,000.
neuropeptide hormone; plays a role in the responses to stress and pain perception.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: enkephalin; preproenkephalin 1; preproenkephalin 2; proenkephalin 1; proenkephalin 2; proenkephalin related sequence; Proenkephalin-A; spermatogenic-specific proenkephalin
Gene Aliases: AI326464; ENK; Penk; Penk-rs; Penk1; Penk2; PPA
UniProt ID: (Rat) P04094, (Mouse) P22005
Entrez Gene ID: (Rat) 29237, (Mouse) 18619
Molecular Function:
neuropeptide
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support