Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
Immunogen Sequence: ARHLQPGMVL TVEPGIYFID HLLDEALADP ARASFLNREV LQRFRGFGGV RIEEDVVVTD SGIELLTCVP RTVEEIEACM AGCDKAFTPF SGPK
Xaa-Pro dipeptidase is a cytosolic dipeptidase that hydrolyzes dipeptides with proline or hydroxyproline at the carboxy terminus (but not Pro-Pro). It is important in collagen metabolism because of the high levels of iminoacids. Xaa-Pro dipeptidase is a cytosolic dipeptidase that hydrolyzes dipeptides with proline or hydroxyproline at the carboxy terminus (but not Pro-Pro). It is important in collagen metabolism because of the high levels of iminoacids.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: aminoacyl-L-proline hydrolase; Imidodipeptidase; MGC10905; Peptidase 4; Peptidase D; Prolidase; Proline dipeptidase; testicular tissue protein Li 138; X-Pro dipeptidase; Xaa-Pro dipeptidase
Gene Aliases: dal; Pep-4; Pep4; PEPD; PRD; PROLIDASE
UniProt ID: (Human) P12955, (Mouse) Q11136
Entrez Gene ID: (Human) 5184, (Mouse) 18624
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support