Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
PER2, also known as Period circadian protein homolog 2, is a mainly nuclear protein that shows nucleocytoplasmic shuttling which is effected by interaction with other circadian core oscillator proteins and/or by phosphorylation. PER2 belongs to the basic helix-loop-helix family of transcription factors and contains a PAC (PAS-associated C-terminal) domain and two PAS (PER-ARNT-SIM) domains. PER2 is a component of the circadian core oscillator, which includes the CRY proteins, CLOCK or NPAS2, BMAL1 or BMAL2, CSNK1D and/or CSNK1E, TIMELESS, and the PER proteins. This protein is thus essential for generating circadian rhythms and is a negative element in the circadian transcriptional loop which influences clock function by interacting with other circadian regulatory proteins and transporting them to the nucleus. Expression of PER2 is fairly wide spread with strong expression in skeletal muscle and pancreas and slight expression in lung. Defects in PER2 are a cause of familial advanced sleep-phase syndrome (FASPS).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Circadian clock protein PERIOD 2; hPER2; mPER2; per2 {ECO:0000312|RGD:61945}; period 2; period circadian protein 2; Period circadian protein homolog 2; period homolog 2; rPER2
Gene Aliases: FASPS; FASPS1; KIAA0347; mKIAA0347; mPer2; PER2; rPER2
UniProt ID: (Human) O15055, (Mouse) O54943, (Rat) Q9Z301
Entrez Gene ID: (Human) 8864, (Mouse) 18627, (Rat) 63840
Molecular Function:
transcription cofactor
gene-specific transcriptional regulator
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support