Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MDAWVRFSAQ SQARERLCRA AQYACSLLGH ALQRHGASPE LQKQIRQLES HLSLGRKLLR LGNSADALES AKRAVHLSDV VLRFCITVSH LNRALYFA
Peroxisomal membrane protein 11B is a protein that in humans is encoded by the PEX11B gene. Involved in peroxisomal proliferation. May regulate peroxisomes division by recruiting the dynamin-related GTPase DNM1L to the peroxisomal membrane.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Peroxin 11 beta; Peroxin 11B; Peroxin-11 beta; Peroxin-11B; Peroxin11B; Peroxisomal biogenesis factor 11B; Peroxisomal membrane protein 11B; PEX-11 beta; PEX11 beta; PEX11-beta; Protein PEX11 homolog beta
Gene Aliases: PEX11-BETA; PEX11B; PEX14B
UniProt ID: (Human) O96011
Entrez Gene ID: (Human) 8799
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support