Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Sandwich ELISA (Recombinant protein).
Immunogen sequence: LGPQEEGEGV VDVKGQVRME VQGEEEKRED KEDEEDEEDD DVSHVDEEDC LGVQREDRRG GDGQINEQVE KLRRPEGASN ESE
PEX14 belongs to the peroxin 14 family. It is a component of the peroxisomal translocation machinery with PEX13 and PEX17 and interacts with both the PTS1 and PTS2 receptors. PEX14 binds directly to PEX17. Defects in PEX14 are the cause of peroxisome biogenesis disorder complementation group K (PBD-CGK) and Zellweger syndrome (ZWS).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: NF-E2 associated polypeptide 2; Peroxin 14; Peroxin-14; Peroxisomal membrane anchor protein PEX14; peroxisomal membrane anchor protein Pex14p; Peroxisomal membrane protein PEX14; PEXE; PTS1 receptor docking protein; PTS1 receptor-docking protein
Gene Aliases: dJ734G22.2; NAPP2; PBD13A; PEX14; Pex14p
UniProt ID: (Human) O75381
Entrez Gene ID: (Human) 5195
Molecular Function:
primary active transporter
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support