Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MAAAEEGCS VGAEADRELE ELLESALDDF DKAKPSPAPP STTTAPDASG PQKRSPGDTA KDALFASQEK FFQELFDSEL ASQATAEFEK AMKELAEEEP HLVEQFQKLS EAAGRVGSDM TSQQEFTSCL KETLSGLAKN ATDLQNSSMS EEELTKAMEG LGMDEGDGEG NILPIMQSIM QNLLSKDVLY PSLKEITEKY PEWLQSHRES LPPEQFEKYQ EQHSVMCKIC EQFEAETPTD SETTQKARFE MVLDLMQQLQ DLGHPPKELA GEMPPGLNFD LDALNLSGPP GASGEQCLIM (1-299 aa encoded by BC000496)
This gene is necessary for early peroxisomal biogenesis. It acts both as a cytosolic chaperone and as an import receptor for peroxisomal membrane proteins (PMPs). Peroxins (PEXs) are proteins that are essential for the assembly of functional peroxisomes. The peroxisome biogenesis disorders (PBDs) are a group of genetically heterogeneous autosomal recessive, lethal diseases characterized by multiple defects in peroxisome function. The peroxisomal biogenesis disorders are a heterogeneous group with at least 14 complementation groups and with more than 1 phenotype being observed in cases falling into particular complementation groups. Although the clinical features of PBD patients vary, cells from all PBD patients exhibit a defect in the import of one or more classes of peroxisomal matrix proteins into the organelle. Defects in this gene are a cause Zellweger syndrome (ZWS).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 33 kDa housekeeping protein; housekeeping gene, 33kD; Peroxin-19; Peroxisomal biogenesis factor 19; Peroxisomal farnesylated protein; peroxisome biogenesis factor 19; PxF
Gene Aliases: D1S2223E; HK33; OK/SW-cl.22; PBD12A; PEX19; PMP1; PMPI; PXF; PXMP1
UniProt ID: (Human) P40855, (Mouse) Q8VCI5
Entrez Gene ID: (Human) 5824, (Mouse) 19298
Molecular Function: membrane traffic protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support