Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Cell lysate).
Immunogen sequence: RVLSAITRKF KLEPSVSLVN VLDCCPPQLT GADLYSLCSD AMTAALKRRV HDLEEGLEPG SSALMLTMED LLQAAARLQP SVSEQELLRY KRIQRKFAAC
PEX6 encodes a member of the AAA (ATPases associated with diverse cellular activities) family of ATPases. This member is a predominantly cytoplasmic protein, which plays a direct role in peroxisomal protein import and is required for PTS1 (peroxisomal targeting signal 1, a C-terminal tripeptide of the sequence ser-lys-leu) receptor activity. Mutations in this gene cause peroxisome biogenesis disorders of complementation group 4 and complementation group 6.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: PAF 2; PAF-2; Peroxin 6; Peroxin-6; peroxisomal AAA-type ATPase 1; Peroxisomal ATPase PEX6; Peroxisomal biogenesis factor 6; Peroxisomal-type ATPase 1; Peroxisome assembly factor 2; RP1-125M3.3
Gene Aliases: HMLR2; PAF-2; PAF2; PBD4A; PDB4B; PEX6; PXAAA1
UniProt ID: (Human) Q13608
Entrez Gene ID: (Human) 5190
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support