Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 10
Positive Control: PC-3M, LOVO, SH-SY-5Y, rat brain tissue, human liver cancer tissue, human thyroid gland tissue, mouse skeletal muscle tissue, human kidney tissue.
Tight junctions represent one mode of cell-to-cell adhesion in epithelial or endothelial cell sheets, forming continuous seals around cells and serving as a physical barrier to prevent solutes and water from passing freely through the paracellular space. The protein encoded by this immunoglobulin superfamily gene member is localized in the tight junctions between high endothelial cells. Unlike other proteins in this family, the this protein is unable to adhere to leukocyte cell lines and only forms weak homotypic interactions. The encoded protein is a member of the junctional adhesion molecule protein family and acts as a receptor for another member of this family.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 6-phosphofructo-1-kinase; 6-phosphofructokinase type A; 6-phosphofructokinase, muscle type; ATP-dependent 6-phosphofructokinase, muscle type; ATP-PFK; PFK-A; Phosphofructo-1-kinase isozyme A; phosphofructokinase 1; phosphofructokinase muscle; phosphofructokinase, polypeptide X; phosphofructokinase-M; Phosphohexokinase; protein phosphatase 1, regulatory subunit 122
Gene Aliases: ATP-PFK; GSD7; PFK-1; PFK1; PFKA; PFKM; PFKX; PPP1R122
UniProt ID: (Human) P08237
Entrez Gene ID: (Human) 5213
Molecular Function:
carbohydrate kinase
kinase
transferase
metabolite interconversion enzyme
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support