Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 4
Phosphoglyceric acid mutase is widely distributed in mammalian tissues where it catalyzes the reversible reaction of 3-phosphoglycerate to 2-phosphoglycerate in the glycolytic pathway.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: MGC88743; OTTHUMP00000059414; PGAM2; PGAMA; PGAMM; phosphoglycerate mutase family 3; phosphoglycerate mutase processed protein; Probable phosphoglycerate mutase 4; RP11-452K12.8
Gene Aliases: dJ1000K24.1; PGAM-B; PGAM1; PGAM3; PGAM4
UniProt ID: (Human) Q8N0Y7
Entrez Gene ID: (Human) 441531
Molecular Function: mutase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support