Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Displays phosphatase activity for serine/threonine residues, and, dephosphorylates and activates MAP3K5 kinase. Has apparently no phosphoglycerate mutase activity. May be regulator of mitochondrial dynamics. Substrate for a KEAP1-dependent ubiquitin ligase complex. Contributes to the repression of NFE2L2-dependent gene expression. Acts as a central mediator for programmed necrosis induced by TNF, by reactive oxygen species and by calcium ionophore. [UniProt]
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Bcl-XL-binding protein v68; PGAM family member 5, serine/threonine protein phosphatase, mitochondrial; Phosphoglycerate mutase family member 5; Serine/threonine-protein phosphatase PGAM5, mitochondrial
Gene Aliases: 2610528A17Rik; BXLBV68; PGAM5; RGD1312028
UniProt ID: (Human) Q96HS1, (Rat) Q562B5, (Mouse) Q8BX10
Entrez Gene ID: (Rabbit) 100338700, (Pig) 106508765, (Human) 192111, (Rat) 288731, (Mouse) 72542
Molecular Function:
isomerase
mutase
metabolite interconversion enzyme
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support