Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
The piggyBac family of proteins, found in diverse animals, are transposases related to the transposase of the canonical piggyBac transposon from the moth, Trichoplusia ni. This family also includes genes in several genomes, including human, that appear to have been derived from the piggyBac transposons. This gene belongs to the subfamily of piggyBac transposable element derived (PGBD) genes. The PGBD proteins appear to be novel, with no obvious relationship to other transposases, or other known protein families.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DKFZp761A0620; FLJ11413; piggyBac domain related protein 5; PiggyBac domain-related protein 5; PiggyBac transposable element-derived protein 5; PiggyBac transposase 5
Gene Aliases: PGBD5
UniProt ID: (Human) Q8N414
Entrez Gene ID: (Human) 79605
Molecular Function:
viral or transposable element protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support