Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
This Antibody was verified by Relative expression to ensure that the antibody binds to the antigen stated.
FIGURE: 1 / 1
Immunogen sequence: FNRTVSKVDD FLANEAKGTK VVGAQSLKEM VSKLKKPRRI ILLVKAGQAV DDFIEKLVPL LDTGDIIIDG GNSEYRDTTR RCRDLKAKGI LFVGS
6-phosphogluconate dehydrogenase is the second dehydrogenase in the pentose phosphate shunt. Deficiency of this enzyme is generally asymptomatic, and the inheritance of this disorder is autosomal dominant. Hemolysis results from combined deficiency of 6-phosphogluconate dehydrogenase and 6-phosphogluconolactonase suggesting a synergism of the two enzymopathies.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 6-phosphogluconate dehydrogenase, decarboxylating
Gene Aliases: 0610042A05Rik; 6PGD; AU019875; C78335; PGD; PGDH
UniProt ID: (Human) P52209, (Mouse) Q9DCD0
Entrez Gene ID: (Human) 5226, (Mouse) 110208
Molecular Function:
dehydrogenase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support