Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Neutralization: At a concentration of 0.05-0.15 µg/mL, this will block 50% of the binding of recombinant mouse PlGF2 (4 ng/mL) to immobilized recombinant human Flt1/Fc Chimera (4 µg/mL of a 100 µL solution coated in each well) in an ELISA.
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Placental growth factor (PlGF) is a member of the vascular endothelial growth factor (VEGF) family. PlGF is a pleiotropic factor affects different cell types and regulates various biological responses. The activity of vascular endothelial growth factor (VEGF), which is the most potent of all angiogenic mediators, is partly modulated by PlGF. It plays a major role in vasculogenesis and angiogenesis in human placenta.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Placenta growth factor; PlGF; PlGF 1; SHGC
Gene Aliases: AI854365; Pgf; PIGF; Plgf
UniProt ID: (Mouse) P49764
Entrez Gene ID: (Mouse) 18654
Molecular Function:
growth factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support