Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Sequence of this protein is as follows: MKGQIRVAKR RVVMASLYLG TGPLEQELVD CLESTLEKSL QAKFPSNLKV SILLDFTRGS RGRKNSRTML LPLLRRFPEQ VRVSLFHTPH LRGLLRLLIP ERFNETIGLQ HIKVYLFDNS VILSGANLSD SYFTNRQDRY VFLQDCAEIA DFFTELVDAV GDVSLQLQGD DTVQVVDGMV HPYKGDRAEY CKAANKRVMD VINSARTRQQ MLHAQTFHSN SLLTQEDAAA AGDRRPAPDT WIYPLIQMKP FEIQIDEIVT ETLLTEAERG AKVYLTTGYF NLTQAYMDLV LGTRAEYQIL LASPEVNGFF GAKGVAGAIP AAYVHIERQF FSEVCSLGQQ ERVQLQEYWR RGWTFHAKGL WLYLAGSSLP CLTLIGSPNF GYRSVHRDLE AQIAIVTENQ ALQQQLHQEQ EQLYLRSGVV SSATFEQPSR QVKLWVKMVT PLIKNFF
PGS1 functions in the biosynthesis of the anionic phospholipids phosphatidylglycerol and cardiolipin.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CDP-diacylglycerol--glycerol-3-phosphate 3-phosphatidyltransferase, mitochondrial; PGP synthase 1; Phosphatidylglycerophosphate synthase 1
Gene Aliases: PGS1
UniProt ID: (Human) Q32NB8
Entrez Gene ID: (Human) 9489
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support