Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
PHLPP1 is a member of the serine/threonine phosphatase family, which are important regulators of Akt serine-threonine kinases (AKT1, AKT2, AKT3) and conventional/novel protein kinase C (PKC) isoforms. PHLPP1 and PHLPP2 have a similar domain structure and have been shown to dephosphorylate and inactivate, distinct Akt isoforms, at one of the two critical phosphorylation sites required for activation: Serine473. PHLPP2 dephosphorylates AKT1 and AKT3, whereas PHLPP1 is specific for AKT2 and AKT3. PHLPP1 promotes apoptosis and may act as a tumor suppressor. Increased expression of PHLPP1 may also play a role in obesity and type 2 diabetes by interfering with Akt-mediated insulin signaling.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: EC 3.1.3.16; hSCOP; PH domain leucine-rich repeat-containing protein phosphatase 1; PH domain-containing family E member 1; pleckstrin homology domain containing, family E (with leucine rich repeats) member 1; Pleckstrin homology domain-containing family E member 1; protein phosphatase, Mg2+/Mn2+ dependent 3A; SCN circadian oscillatory protein; Suprachiasmatic nucleus circadian oscillatory protein
Gene Aliases: KIAA0606; PHLPP; PHLPP1; PLEKHE1; PPM3A; SCOP
UniProt ID: (Human) O60346
Entrez Gene ID: (Human) 23239
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support