Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 4
Immunogen sequence: MEMGRRIHLE LRNRTPSDVK ELVLDNSRSN EGKLEGLTDE FEELEFLSTI NVGLTSIANL PKLNKLKKLE LSDNRVSGGL EVLAEKCPNL THLNLSGNKI KDLSTIEPLK KLENLKSLDL FNCEVTNLND YRENVFKLLP QLTYLDGYDR DDKEAPDSDA EGYVEGLDDE; Positive Samples: 293T, HepG2, HL-60, MCF7, Raji, U-251MG, Mouse brain, HT-29; Cellular Location: Cytoplasm, Endoplasmic reticulum, Nucleus
PHAP I is implicated in a number of cellular processes, including proliferation, differentiation, caspase-dependent and caspase-independent apoptosis, suppression of transformation (tumor suppressor), inhibition of protein phosphatase 2A, regulation of mRNA trafficking and stability in association with ELAVL1, and inhibition of acetyltransferases as part of the INHAT (inhibitor of histone acetyltransferases) complex. Plays a role in E4F1-mediated transcriptional repression.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Acid nuclear phosphoprotein 32 (leucine rich); acidic (leucine-rich) nuclear phosphoprotein 32 family, member A; Acidic leucine-rich nuclear phosphoprotein 32 family member A; acidic nuclear phosphoprotein 32; acidic nuclear phosphoprotein 32 family, member A; Acidic nuclear phosphoprotein pp32; cerebellar leucine rich acidic nuclear protein; hepatopoietin Cn; inhibitor-1 of protein phosphatase-2A; LANP; Leucine-rich acidic nuclear protein; leucine-rich acidic nuclear protein LANP; Mapmodulin; MGC119787; MGC150373; PHAPI; Potent heat-stable protein phosphatase 2A inhibitor I1PP2A; pp32; Putative HLA-DR-associated protein I; putative human HLA class II associated protein I; vacidic (leucine-rich) nuclear phosphoprotein 32 family, member A
Gene Aliases: Anp32; ANP32A; C15orf1; HPPCn; I1PP2A; LANP; MAPM; PHAP1; PHAPI; PP32; W91701
UniProt ID: (Human) P39687, (Rat) P49911, (Mouse) O35381
Entrez Gene ID: (Human) 8125, (Rat) 25379, (Mouse) 11737
Molecular Function:
chromatin/chromatin-binding, or -regulatory protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support