Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
Promotions
View available promotion(s)
Promo Code: P5906365 Don't miss out: buy 3, only pay for 2 Extended to June 27: take advantage of our 3 for 2 promotion. Whether you are restocking essentials or trying something new, now is a great time to get more for your money!. Learn more
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
Immunogen sequence: RINARLQQY RAKAELARST RPQAWVPREK LPRPLTSSAS AIRKLMRKAE LMGISTDIFP VDNSDTSSSV DGRRKHKQPA LTADFVNYYF ERNMRMIQIQ ENMAEQKNIK DKLENEQEKL HVEYNKLCES LEELQNLNGK LRSEGQGIWA LLGRITGQKL NIPAILRAPK ERKPSKKEGG TQKTSTLPAV LYSCGICKKN HDQHLLLLCD TCKLHYHLGC LDPPLTRMPR KTKNSYWQCS ECDQAGSSDM EADMAMETLP DGTKRSRRQI KEPVKFVPQD VPPEPKKIPI RNTRTRGRKR SFVPEEEKHE ERVPRERRQR QSVLQKKPKA EDLRTECATC KGTGDNENLV R (536-885 aa encoded by BC152414)
PHF14 gene ontology annotations related to this gene include metal ion binding.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: PHD finger protein 14
Gene Aliases: KIAA0783; PHF14
UniProt ID: (Human) O94880
Entrez Gene ID: (Human) 9678
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support