Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MAFANLRKVL ISDSLDPCCR KILQDGGLQV VEKQNLSKEE LIAELQDCEG LIVRSATKVT ADVINAAEKL QVVGRAGTGV DNVDLEAATR KGILVMNTPN GNSLSAAELT CGMIMCLARQ IPQATASMKD GKWERKKFMG TELNGKTLGI LGLGRIGREV ATRMQSFGMK TIGYDPIISP EVSASFGVQQ LPLEEIWPLC DFITVHTPLL PSTTGLLNDN TFAQCKKGVR VVNCARGGIV DEGALLRALQ SGQCAGAALD VFTEEPPRDR ALVDHENVIS CPHLGASTKE AQSRCGEEIA VQFVDMVKGK SLTGVVNAQA LTSAFSPHTK PWIGLAEALG TLMRAWAGSP KGTIQVITQG TSLKNAGNCL SPAVIVGLLK EASKQADVNL VNAKLLVKEA GLNVTTSHSP AAPGEQGFGE CLLAVALAGA PYQAVGLVQG TTPVLQGLNG AVFRPEVPLR RDLPLLLFRT QTSDPAMLPT MIGLLAEAGV RLLSYQTSLV SDGETWHVMG ISSLLPSLEA WKQHVTEAFQ FHF
PHGDH gene encodes the enzyme which is involved in the early steps of L-serine synthesis in animal cells. L-serine is required for D-serine and other amino acid synthesis. The enzyme requires NAD/NADH as a cofactor and forms homotetramers for activity. Mutations in this gene have been found in a family with congenital microcephaly, psychomotor retardation and other symptoms. Multiple alternatively spliced transcript variants have been found, however the full-length nature of most are not known.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 2-oxoglutarate reductase; 3-PGDH; 3-phosphoglycerate dehydrogenase; D-3-phosphoglycerate dehydrogenase; epididymis secretory protein Li 113; Malate dehydrogenase
Gene Aliases: 3-PGDH; 3PGDH; HEL-S-113; NLS; NLS1; PDG; PGAD; PGD; PGDH; PGDH3; PHGDH; PHGDHD; SERA
UniProt ID: (Human) O43175
Entrez Gene ID: (Human) 26227
Molecular Function:
dehydrogenase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support