Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Sequence of this protein is as follows: MTRDEALPDS HSAQDFYENY EPKEILGRGV SSVVRRCIHK PTSQEYAVKV IDVTGGGSFS PEEVRELREA TLKEVDILRK VSGHPNIIQL KDTYETNTFF FLVFDLMKRG ELFDYLTEKV TLSEKETRKI MRALLEVICT LHKLNIVHRD LKPENILLDD NMNIKLTDFG FSCQLEPGER LREVCGTPSY LAPEIIECSM NEDHPGYGKE VDMWSTGVIM YTLLAGSPPF WHRKQMLMLR MIMSGNYQFG SPEWDDYSDT VKDLVSRFLV VQPQNRYTAE EALAHPFFQQ YLVEEVRHFS PRGKFKVIAL TVLASVRIYY QYRRVKPVTR EIVIRDPYAL RPLRRLIDAY AFRIYGHWVK KGQQQNRAAL FENTPKAVLL SLAEEDY
PHKG1 is a serine/threonine kinase with glycogenolytic regulatory functions. This protein is the catalytic member of a 16 subunit protein kinase complex which contains equimolar ratios of 4 subunit types. The complex is a crucial glycogenolytic regulatory enzyme. PHKG1 has two pseudogenes at chromosome 7q11.21 and one at chromosome 11p11.12.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: PHK-gamma-M; PHK-gammaM; PHKgamma-M; PHKgammaM; phosphorylase b kinase gamma catalytic chain, skeletal muscle isoform; Phosphorylase b kinase gamma catalytic chain, skeletal muscle/heart isoform; phosphorylase kinase; phosphorylase kinase gamma; phosphorylase kinase gamma subunit 1; Phosphorylase kinase subunit gamma-1; phosphorylase kinase, gamma 1 (muscle); Serine/threonine-protein kinase PHKG1
Gene Aliases: PHKG; PHKG1
UniProt ID: (Human) Q16816
Entrez Gene ID: (Human) 5260
Molecular Function:
non-receptor serine/threonine protein kinase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support